missing translation for 'onlineSavingsMsg'
Learn More

Ibandronate Sodium Monohydrate, Mikromol™

Product Code. 17961276
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17961276

Brand: Mikromol MM1326.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ibandronate Sodium Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibandronate Sodium Monohydrate
CAS 138926-19-9
Analyte or Component Names Ibandronate Sodium Monohydrate
Shipping Condition Room Temperature
Molecular Formula C9 H22 N O7 P2 . Na . H2 O
Certifications/Compliance ISO 17034
Synonym Ibandronate Sodium Monohydrate, P,P'-[1-hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid sodium salt hydrate (1:1:1), [1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid monosodium salt monohydrate, BM 21.0955Na.H2O, Ibandronate sodium monohydrate, Monosodium ibandronate monohydrate
InChI Formula InChI=1S/C9H23NO7P2.Na.H2O/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17;;/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17);;1H2/q;+1;/p-1
SMILES O.[Na+].CCCCCN(C)CCC(O)(P(=O)(O)O)P(=O)(O)[O-]
IUPAC Name sodium;hydroxy-[1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl]phosphinate;hydrate
Molecular Weight (g/mol) 359.23
Quantity 250 mg
Formula Weight 359.0875 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.