missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibandronate Sodium Monohydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ibandronate Sodium Monohydrate |
| CAS | 138926-19-9 |
| Analyte or Component Names | Ibandronate Sodium Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H22 N O7 P2 . Na . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ibandronate Sodium Monohydrate, P,P'-[1-hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid sodium salt hydrate (1:1:1), [1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid monosodium salt monohydrate, BM 21.0955Na.H2O, Ibandronate sodium monohydrate, Monosodium ibandronate monohydrate |
| InChI Formula | InChI=1S/C9H23NO7P2.Na.H2O/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17;;/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17);;1H2/q;+1;/p-1 |
| Solution Type | Neat |
| SMILES | O.[Na+].CCCCCN(C)CCC(O)(P(=O)(O)O)P(=O)(O)[O-] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction