missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ siRNA Labeling Kit with FAM dye
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
For the labeling of siRNA with FAM™. Includes sufficient reagents for labeling 65μg of siRNA. Invitrogen™ Labeled siRNA can be used to analyze siRNA subcellular localization, stability, and transfection efficiency. In addition, fluorescently labeled siRNA is particularly well-suited for use in double-label experiments (with a labeled antibody) to track cells that receive siRNA during transfection and to correlate transfection with down-regulation of the target protein. Research shows that labeling of siRNAs does not affect their biological function.
Accessory Products:
Also available is the Silencer™ Cy™3 siRNA Labeling Kit (SKU# AM1632).
Accessory Products:
Also available is the Silencer™ Cy™3 siRNA Labeling Kit (SKU# AM1632).
Specifications
Specifications
| Content And Storage | 10X Labeling Buffer, Reconstitution Solution, 5M NaCl, 5X siRNA Annealing Buffer, GAPDH siRNA, and FAM™ Labeling Reagent should be stored at –20°C. Nuclease-free water may be stored at any temperature. |
| Detection Method | Fluorescence |
| Format | Kit |
| Label or Dye | FITC |
| Final Product Type | siRNA (Labeled) |
| Labeling Target | siRNA |
| Includes Label or Dye | Yes |
| Labeling Method | Direct Labeling |
| Product Line | Silencer, Ambion |
| Product Type | siRNA Labeling Kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction