Learn More
Masterflex™ I/P™ High-Performance Pump Head Mounting Screws
Product Code. 15497082
Change view
Click to view available options
For Use With:
77600-62 Pump Head
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15497082 77600-62 Pump Head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15497082 Supplier Masterflex™ Supplier No. 7760001
Replacement parts to keep your Masterflex™ I/P™ pump heads reliable
  • Replacement mounting screws for the 77600-62 pump head
  • Mounts head to I/P drive
Specifications
For Use With (Application) Mounting Pump Head to I/P Drive
For Use With 77600-62 Pump Head