Learn More
I-BET151, MedChemExpress
Product Code. 16491737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491737 50 mg 50mg
16425578 10 mg 10mg
16408277 5 mg 5mg
16406126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16491737 Supplier Medchem Express Supplier No. HY13235
MedChemExpress I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.
I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.
Specifications
Chemical Name or Material I-BET151
CAS 1300031-49-5
Color White
Purity Grade Notes Research
Molecular Formula C23H21N5O3
Quantity 50 mg
Synonym GSK1210151A
Solubility Information DMSO : ≥ 100 mg/mL (240.71 mM)
SMILES C[C@@H](N1C(C(C=C(OC)C(C2=C(C)ON=C2C)=C3)=C3N=C4)=C4NC1=O)C5=CC=CC=N5
Molecular Weight (g/mol) 415.44
Formula Weight 415.44
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death