missing translation for 'onlineSavingsMsg'
Learn More

I-BET151, MedChemExpress

Product Code. 16408277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408277 5 mg 5mg
16491737 50 mg 50mg
16425578 10 mg 10mg
16406126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16408277 Supplier Medchem Express Supplier No. HY13235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.

I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.

Specifications

Chemical Name or Material I-BET151
CAS 1300031-49-5
Color White
Purity Grade Notes Research
Molecular Formula C23H21N5O3
Quantity 5 mg
Synonym GSK1210151A
Solubility Information DMSO : ≥ 100 mg/mL (240.71 mM)
SMILES C[C@@H](N1C(C(C=C(OC)C(C2=C(C)ON=C2C)=C3)=C3N=C4)=C4NC1=O)C5=CC=CC=N5
Molecular Weight (g/mol) 415.44
Formula Weight 415.44
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.