Learn More
Thermo Scientific™ Hypersil™ BDS C8 Javelin™ Guard Columns
Product Code. p-3134048 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
2.1 mm
Length (Metric):
20 mm
Unit Size:
Pack of 4
Product Code. 11573161
Brand: Thermo Scientific™ 28205022106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offer convenient, easy-to-use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of BDS C8 Solid Phase
  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C8