Learn More
Thermo Scientific™ Hypersil™ APS-2 Amino HPLC Columns
Product Code. 16338586 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
20 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 4
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
16338586 250 mm 20 mm 5 μm Each
10670916 50 mm 2.1 mm 3 μm Each
10553045 50 mm 4.6 mm 3 μm Each
10007174 100 mm 2.1 mm 3 μm Each
10495913 150 mm 2.1 mm 3 μm Each
10405923 150 mm 4.6 mm 3 μm Each
10755973 250 mm 4.6 mm 5 μm Each
10414393 150 mm 4.6 mm 5 μm Each
10039642 100 mm 4.6 mm 5 μm Each
11563041 100 mm 3 mm 5 μm Each
10631514 10 mm 4 mm 5 μm Pack of 4
10107894 10 mm 3 mm 5 μm Pack of 4
10269314 10 mm 4 mm 3 μm Pack of 4
10774047 10 mm 2.1 mm 3 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 16338586 Fournisseur Thermo Scientific™ Code fournisseur 30705259270A
Il en reste null
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns.
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns. Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines. For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ LC Amino Columns as an alternative.
  • In reversed phase mode, Hypersil APS-2 columns are excellent for carbohydrate analysis: extra sensitivity for sugars
  • Alternative selectivity to silica when used in normal phase
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Spécification
Column Format Preparative Column, Standard
Diameter (Metric) 20 mm
For Use With Preparative HPLC
Length (Metric) 250 mm
Max. Pressure 4300 psi (295 bar)
Particle Size 5 μm
pH 2.5 to 8
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L8
Stationary Phase Amino (NH2)
Packing Material Spherical, Fully Porous Type A Silica
Carbon Load 2%
Column Type Carbohydrate Analysis
Endcapped No
Phase Reversed Phase, Normal Phase, Ion Exchange, HILIC
Afficher plus Afficher moins
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.