Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Empty Well Plates
Shop All Thermo Scientific Products
Click to view available options
Column Capacity (Metric):
1 mL
2 mL
Unit Size:
Each
Pack of 40
Description
- HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format.
- Available in a wide range of chemistries and bed weights
- Ideal for large samples and conform to industry standard configurations
- Allows for efficient routine processing by vacuum, by positive pressure or centrifugation.
- The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility.
- The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
- Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
- HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Specifications
| Column Capacity (Metric) | 1 mL |
| For Use With (Application) | Solid Phase Extraction |
| Type | 96-well SPE Plate |