Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Shop All Thermo Scientific ProductsDescription
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
Comprehensive range of Dispersive SPE products that contain the proper sorbents foroptimum extraction, clean-up and separation of analytes from complex matrices such as food using QuEChERS methods.
- Save valuable time in preparation – products are supplied pre-prepared
- Reduced variability due to consistency in products and rigorous quality control
- Cleaner extracts from cleaner, pre-prepared products
- Lower peak counts as a result of using pre-prepared products
Specifications
Specifications
| Description | 900 mg anhydrous magnesium sulfate, 300 mg PSA |
| For Use With (Application) | QuEChERS Method |
| Product Line | Quechers |
| Sorbent | 900 mg MgSO4, 300 mg PSA |
| Volume (Metric) | 15 mL |
| Quantity | 50 |