Learn More
Thermo Scientific™ HyPerforma™ Mixtainer™ Hardware, Stainless-Steel Support Plate; EU
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific™ Stain-less Steel Support Plate (EU) is part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a conical drum, a magnetic stirring plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.
Recommended for:
- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Specifications
Specifications
| Dimensions | 14.19 x 14.19 x 4 in. |
| Quantity | Each |
| Material | Stainless Steel (304) |
| Product Line | HyPerforma, Mixtainer |