Learn More
Ansell HyFlex™ 11-738 Intercept™ Cut-Resistant Gloves with Targeted Reinforcement
GREENER_CHOICE
Product Code. 16009661 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16009661
Brand: Ansell 11738070
Fortix™ Technology increases protection with additional durability in the thumb crotch region, and extends the glove's useful life.
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology extends product's useful life
  • Increased protection in the thumb crotch region
  • Water-based polyurethane enhances comfort and dexterity
  • Oeko-Tek™ 100 Certified
  • Solvent-free
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 4X42C
For Use With (Application) Automotive Assembly, Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-738
Yarn Gauge 13