Learn More
Ansell™ HyFlex™ 11-616 Series Black and Green/Blue Polyurethane Light Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Description
- 18-gauge knitting technology allows this glove to be ultra-thin yet tough
- Ideal for workers who need a higher level of fit, touch and precision
- Ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries.
Specifications
Specifications
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, EN 388 : 3121 |
| Cuff Material | Polyurethane |
| Coating Material | Polyurethane |
| Color | Blue |
| Exterior Finish | No Texture |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nylon |
| Product Type | Lightweight Gloves |
| Show More |