missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-515 Hi-Vis Cut-Resistant Nitrile Gloves with Intercept Technology™ Construction

Product Code. 16039601 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16039601

Brand: Ansell 11515090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Intercept Technology™ yarn incorporates DuPont™ Aramid Fiber Stretch Armor for ANSI Level 5 Cut Performance. Gloves are ergonomically designed with enhanced grip capabilities in dry or light oil applications.

  • Gloves combine comfort and durability with ANSI Level 5 Cut Performance
  • Ergonomically designed with hi-vis colors to better protect users in low light environments
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Extended cuff provides additional cut protection in the highly sensitive wrist area
  • Foam nitrile formulation provides longer wear for increased durability
  • Enhances grip and tactility in light oil or dry environments for grip assurance when handling sharp objects and knives
  • Provides workers with the ability to perform safely and efficiently
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coating Material Nitrile
Color Orange
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 4X42E
For Use With (Application) Automotive Assembly, Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Series 11-515
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.