missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ 11-280 Sleeves
GREENER_CHOICE

Product Code. 17647455
Click to view available options
Size:
Narrow
Wide
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 17647455

Brand: Ansell™ 11280160N

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-280 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort

  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
TRUSTED_SUSTAINABILITY

Specifications

Size Narrow
Construction Knitted
Product Type Sleeve
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Latex Free Yes
Model HyFlex™11-280
For Use With (Application) Glass repair and replacement; Handling metal sheets and panels
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Length (Metric) 40 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.