Learn More
Ansell™ HyFlex™ 11-100
Product Code. 16059881 Shop All Ansell Products
Click to view available options
Size:
6
7
Unit Size:
12 pairs
Pack of 12
Product Code. 16059881
Brand: Ansell™ 11100070
Only null left
Add to basket
Add to basket
Outstanding comfort and product protection from ESD
  • The HyFlex™ 11-100 gloves are especially designed for micro-electronic work where electro-static discharge (ESD) must be avoided.
  • The thin design improves the handling of ever smaller components.
  • Ansell is unique in offering a nitrile foam version. The HyFlex™ 11-100 gloves with nitrile foam ensure that surfaces stay clean and uncontaminated.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Conductive fiber, Nylon
Certifications/Compliance EN388:2016/2131A
Classification Category II
Coated Area Palm coated
Cuff Material Knitwrist
Description Outstanding comfort and product protection from ESD
Coating Material Nitrile Rubber
Color Gray
Cut Resistance Rating ISO Cut Level A
Disposable Reusable
Exterior Finish Conductive Fiber, Nylon
Grade Industrial grade
Laboratory/Cleanroom Use Contolled-environment rooms
Latex Free Yes
Material Nylon
Powdered Powder Free
Product Type Mechanical Protection Gloves, Special Purpose, Light Duty
Size 7
Sterility Non-sterile
Style 11-100
Thickness (Metric) 1.09 mm
For Use With (Application) Mechanical Protection
Glove Cuff Color Gray
Glove Cuff Style Knitwrist
Hem Color Gray
Liner Material Conductive fiber, Nylon
Lining Knit Lining
Model 11-100
Packaging Type Polybag/cardboard box
Palm Type Palm coated, Ansell GRIP Technology
Puncture Rating Level 1
Resistant To 2131A
Series Hyflex 11 series
Tear Rating Level 3
Dimensions (L x W) 207 x 80 mm
Length (English) 220 mm
Weight (Metric) 15.88 g