missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyfasudil hydrochloride, MedChemExpress

Product Code. p-200097712
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485908

Brand: Medchem Express HY13911A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.

Hydroxyfasudil hydrochloride is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.

Specifications

Chemical Name or Material Hydroxyfasudil hydrochloride
CAS 155558-32-0
Color White
Purity Grade Notes Research
Molecular Formula C14H18ClN3O3S
Quantity 10 mg
Synonym HA-1100 hydrochloride HA 1100 hydrochloride HA1100 hydrochloride
Solubility Information DMSO : 30 mg/mL (87.25 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (9.69 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1NC=CC2=C1C=CC=C2S(=O)(N3CCNCCC3)=O.[H]Cl
Molecular Weight (g/mol) 343.83
Formula Weight 343.83
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.