missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxychloroquine O-Sulfate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxychloroquine O-Sulfate Sodium Salt |
| Analyte or Component Names | Hydroxychloroquine O-Sulfate Sodium Salt |
| Purity Grade Notes | HPLC |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18H25ClN3NaO4S |
| Synonym | 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol 1-(Hydrogen Sulfate) Sodium Salt, 2-[[4-[(7-Chloro-4-quinolyl)amino]pentyl]ethylamino]ethanol Sulfate Sodium Salt |
| InChI Formula | InChI=1S/C18H26ClN3O4S.Na/c1-3-22(11-12-26-27(23,24)25)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;/h6-9,13-14H,3-5,10-12H2,1-2H3,(H,20,21)(H,23,24,25);/q;+1/p-1 |
| Solution Type | Neat |
| SMILES | ClC1=CC2=C(C(NC(C)CCCN(CC)CCOS(=O)(O[Na])=O)=CC=N2)C=C1 |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?