Learn More
VEE GEE Hydrometer Jar
Click to view available options
Capacity (English):
17 fl. oz.
20.3 fl. oz.
3.4 fl. oz.
34 fl. oz.
8.5 fl. oz.
Height (English):
7.3 in.
11 in.
14 in.
15.4 in.
16 in.
Unit Size:
Each
Description
- The capacity varies depending on the model.
Specifications
Specifications
| Base Type | Flat Circular |
| Capacity (English) | 17 fl. oz. |
| Height (English) | 14 in. |
| Height (Metric) | 36 cm |
| Type | Hydrometer Jar |
| Diameter (Metric) | 10.8 cm |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate glass |
| Disposable | Reusable |