Learn More
Hydrogen Peroxide Solution, Contains inhibitor, Honeywell™
Product Code. 15687900 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly bottle
Unit Size:
1 L
500mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15687900
Brand: Honeywell Research Chemicals H3410500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Meets USP testing specifications, contains inhibitor, 30 wt. % in H2O
  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
Specifications
Color Colorless
UN Number UN2014
Beilstein 3587191
Density 1.11g/mL at 25°C
Solubility Information Completely miscible in water
Formula Weight 34.01g/mol
Odor Stinging
Packaging Poly bottle
Vapor Pressure 23.3mmHg (30°C)
pH 2.0 to 4.0 (at 20°C)
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen peroxide solution, Contains inhibitor, 30wt. % in H2O, meets USP testing specifications