Learn More
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Product Code. p-213406 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
Unit Size:
500mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15686804
Brand: Fisher Chemical H325500
Request Bulk or Custom Format
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Hydrogen Peroxide, 30% (Certified ACS)
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
Specifications
Color Colorless
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 29.0 to 32.0 %
Density 1.110g/cm³
Formula Weight 34
Percent Purity 29.0 to 32.0%
Grade ACS
Identification Pass test
Packaging Poly Bottle
Residue after Evaporation 0.002 % max.
Vapor Pressure 23mmHg at 30°C
Decomposition Information Decomposes information: >125°C
pH 3.3
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 30%