Learn More
Hydrochloric Acid Solution, 18% by Weight, Chem-Lab
Product Code. 11973004
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. 11973004
Brand: Chem-Lab CL02.0319.5000
Used as a laboratory reagent and acidifying agent. Hydrochloric Acid Solution, 18% by Weight, Chem-Lab contains 202g HCl per liter of water. It is used for pH control, titrations, the production of organic and inorganic compounds, and in the regeneration of ion exchange resins.
Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.
Specifications
Appearance Colorless
pH <1
Color Colorless
CAS 7647-01-0, 7732-18-5
Composition 202g HCl per 1L H2O
Melting Point -25°C
Boiling Point 108°C
Concentration 18% by weight
Density 1.09g/mL
Quantity 5 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Soluble in Water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 7732-18-5 (water)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 18.5 weight % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid