Learn More
Hydrochloric Acid, 0.1M, Chem Lab™
Product Code. 11933094
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11933094 1 L 1 L
11953113 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11933094 Supplier Chem-Lab Supplier No. CL05.0320.1000
Specifications
Appearance Colorless
pH <2
Composition Hydrochloric Acid 0.1mol/L Propanol-2
Concentration 3.646g HCl/l C3H8O = 0.1 N (±0.0002/20°C)
Density 0.79g/mL
Odor 2-Propanol
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 0.1 mol/l in Propanol-2
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid