Learn More
WTW™ Hydrazine Reagent Test Kits
Product Code. 10470653
Change view
Click to view available options
No. of Tests:
100
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10470653 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10470653 Supplier WTW™ Supplier No. 250493
With certificate for optimum precision.
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For low concentrations and routine work with large sample throughput in the laboratory
  • Wide measuring range using rectangular cuvettes for determination of trace concentrations
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For waste water, drinking water, boiler feed water
For Use With (Equipment) For PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 100
Range 0.005 - 2.00mg/L
Test Method Photometric
Type Photoflex™ Turb/set
compliance-icons
Product Identifier
  • Hydrazine Test Method: photometric 0.005 - 2.00 mg/l N
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid