Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Beskrivelse
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Tekniske data
Tekniske data
| Chemical Name or Material | HYDRANAL™-Coulomat AF 7 |
| CAS | 288-32-4, 7446-09-5, 67-56-1 |
| Quantity | 1 L |
| Flash Point | 11°C (51.8°F) |
| Packaging | Glass bottle |
| UN Number | UN1230 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion