Learn More
HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™
Product Code. p-7072139 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
Product Code. 15614480
Brand: Honeywell-Hydranal 34816500ML
Request Bulk or Custom Format
Specifications
Chemical Name or Material HYDRANAL™-Composite 5 K
CAS 288-32-4, 7446-09-5, 7553-56-2, 111-90-0, 68007-08-9
Density 1.15g/mL
Quantity 500 mL
Flash Point 90°C (194°F)
compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, iodine, imidazole, 2-Methylimidazole, sulphur dioxide, 1H-Imidazole monoh