Learn More
Cytiva HyClone™ SG 200 solution
Product Code. 12310033
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12310033 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12310033 Supplier Cytiva Supplier No. SH30590.01
Stable dipeptide of L-Alanyl L-Glutamine which acts as nutrient media supplement.
  • Used as nutrient media supplement
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance cGMP and ISO 91:2 Guidelines
Content And Storage 2° to 8°C
Description SG-200, (Stable dipeptide of L-AlanyL-Glutamine) 200mM, in 0.85% NaCl Solution
Concentration 2mM
Sterilization Method 0.1 μm Sterile-filtered
Endotoxin Level =1.0EU/mL
Quantity 100 mL