missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-12 beta protein

Product Code. p-7225807 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17650087

Brand: Proteintech HZ13211000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ recombinant protein

IL-12p40, also known as IL-12B, is a subunit of both the IL-12 and IL-23 cytokines. It is not thought to be bioactive on its own, but it has been shown to regulate IL-12 activity in antagonistic manner by competitively binding with the IL-12 receptor. IL-12p40 also functions as a chemoattractant molecule for macrophage infiltration into tumor sites. There has additionally been evidence to suggest that IL-12p40 mediates a protective immune response against mycobacterial infection by serving as a necessary component for granuloma formation and promoting Th1 effector cell functions (PMID: 17126601, 10453037, 11739515).

Specifications

Accession Number P29460
Formulation 1 x PBS pH 7.4
Gene ID (Entrez) 3593
Molecular Weight (g/mol) 37-50 kDa
Quantity 1000 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <0.1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.