missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF695 Full-length ORF (AAH23527.1, 1 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16182493
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16182493 25 μg 25µg
16172493 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16182493 Supplier Abnova™ Supplier No. H00057116P02.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MGLLAFRDVALEFSPEEWECLDPAQRSLYRDVMLENYRNLISLGEDSFNMQFLFHSLAMSKPELIICLEARKEPWNVNTEKTAKHSALSSYLTEDILPEQGLQVSFQKVMLRRYERCCLEKLRLRNDWEIVAM

Specifications

Accession Number AAH23527.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57116
Molecular Weight (g/mol) 42.1kDa
Name ZNF695 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MGLLAFRDVALEFSPEEWECLDPAQRSLYRDVMLENYRNLISLGEDSFNMQFLFHSLAMSKPELIICLEARKEPWNVNTEKTAKHSALSSYLTEDILPEQGLQVSFQKVMLRRYERCCLEKLRLRNDWEIVAM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC61907/RP11-551G24.1/SBZF3
Common Name ZNF695
Gene Symbol ZNF695
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.