missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF384 (aa 4-101) Control Fragment Recombinant Protein

Product Code. 30203560
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203560

Brand: Invitrogen™ RP102311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52044 (PA5-52044. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene contains long CAG trinucleotide repeats coding consecutive glutamine residues. The gene product may functions as a transcription factor, with a potential role in the regulation of neurodevelopment or neuroplasticity. The protein appears to bind and regulate the promoters of MMP1, MMP3, MMP7 and COL1A1. Studies in mouse suggest that nuclear matrix transcription factors (NP/NMP4) may be part of a general mechanical pathway that couples cell construction and function during extracellular matrix remodeling. Multiple transcript variants encoding several isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TF68
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 171017
Name Human ZNF384 (aa 4-101) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BB163993; C130073D16Rik; CAG repeat protein 1; CAGH1; CAGH1A; Cas-associated zinc finger protein; Cas-interacting zinc finger protein; Ciz; ERDA2; expanded repeat domain, CAG/CTG 2; NMP4; NP; nuclear matrix protein 4; Nuclear matrix transcription factor 4; TNRC1; Trinucleotide repeat-containing gene 1 protein; Zfp384; Zinc finger protein 384; Znf384
Common Name ZNF384
Gene Symbol ZNF384
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SHFNSNPYFWPSIPTVSGQIENTMFINKMKDQLLPEKGCGLAPPHYPTLLTVPASVSLPSGISMDTESKSDQLTPHSQASVTQNITVVPVPSTGLMTA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.