missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF281 Synthetic Peptide

Product Code. 15225239
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15225239 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15225239 Supplier Invitrogen™ Supplier No. PEP1235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

ZFP281 Synthetic Peptide, PEP-1235, from Invitrogen.

ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, and NANOG. Gene expression microarray anlysis indicated that some ZFP281 target genes were activated, while others were repressed upon knockdown of ZFP281 expression, showing that ZFP281 plays bifuctinal roles in regulating gene expression in stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 23528
Species Human
Purification Method purified
Accession Number Q9Y2X9
Common Name Human ZNF281
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZNF281
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.