Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Base Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |