Learn More
Abnova™ Human VRK3 Partial ORF (NP_001020949, 322 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051231-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Sequence: DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVPSpecifications
NP_001020949 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVP | |
RUO | |
VRK3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51231 | |
VRK3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VRK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |