Learn More
Abnova™ Human VRK3 Partial ORF (NP_001020949, 322 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_001020949 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51231 |
Molecular Weight (g/mol) | 37.07kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025505
|
Abnova™
H00051231-Q01.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16015505
|
Abnova™
H00051231-Q01.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Sequence: DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVPSpecifications
NP_001020949 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VRK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51231 | |
VRK3 (Human) Recombinant Protein (Q01) | |
DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVP | |
RUO | |
VRK3 | |
Recombinant | |
wheat germ expression system |