Learn More
Invitrogen™ Human UBR1 (aa 1194-1311) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q8IWV7 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 197131 |
| Name | Human UBR1 (aa 1194-1311) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI504731; E3 alpha; E3 ubiquitin-protein ligase UBR1; E3a ligase; JBS; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase UBR1; MGC142065; MGC142067; N-recognin-1; RGD1562326; RING-type E3 ubiquitin transferase UBR1; ubiquitin ligase E3 alpha-I; ubiquitin protein ligase E3 component n-recognin 1; ubiquitin-protein ligase e3 componen n-recognin; ubiquitin-protein ligase E3-alpha; ubiquitin-protein ligase E3-alpha-1; Ubiquitin-protein ligase E3-alpha-I; Ubr1 |
| Common Name | UBR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.