missing translation for 'onlineSavingsMsg'
Learn More

Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18769638
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769638 100 μg 100µg
18701727 25 μg 25µg
18724227 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18769638 Supplier R&D Systems Supplier No. MAB210100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 13 publications

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF-alpha
Applications Neutralization
Classification Monoclonal
Clone 1825
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.01-0.06 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01375
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen E. coli-derived recombinant human TNF-alpha Gly57-Leu233 Accession # P01375
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant rhesus macaque TNF-alpha is observed and no cross-reactivity with recombinant cotton rat, mouse, rat, or porcine TNF-alpha is observed. It shows no cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhOX40 Ligand, rhRANK Ligand, rhTNF-beta , rhTRAIL, rhTRANCE, rhTWEAK, or rhVEGI.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.