missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TMEM63B (aa 240-300) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56094 (PA5-56094. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TMEM63B belongs to the SPO75/TMEM63 family. It is a multi-pass membrane protein. The function of TMEM63B remains unknown.
Specifications
Specifications
| Accession Number | Q5T3F8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55362 |
| Name | Human TMEM63B (aa 240-300) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BC026370; C6orf110; CSC1-like protein 2; RGD1305862; RP3-421H19.2; Tmem63b; Tmem63b-ps1; Transmembrane protein 63 B; transmembrane protein 63 B, pseudogene 1; transmembrane protein 63 B; CSC1-like protein 2 |
| Common Name | TMEM63B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction