missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF1487-SP
This item is not returnable.
View return policy
Description
TLR3 Polyclonal specifically detects TLR3 in Human samples. It is validated for Western Blot.
Specifications
| TLR3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7098 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O15455 | |
| TLR3 | |
| Mouse myeloma cell line NS0-derived recombinant human TLR3 Lys27-Ser711 Accession # O15455 | |
| 25 μg | |
| Primary | |
| Detects human TLR3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rhTLR1, rhTLR2, rhTLR4 and rmTLR6 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction