missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tiki1/TRABD2A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8376-SP
This item is not returnable.
View return policy
Description
Tiki1/TRABD2A Monoclonal specifically detects Tiki1/TRABD2A in Human samples. It is validated for Western Blot.
Specifications
| Tiki1/TRABD2A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TRABD2A | |
| Chinese hamster ovary cell line CHO-derived recombinant human Tiki1/TRABD2A Met1-Met477 Accession # Q86V40 | |
| 25 μg | |
| Primary | |
| Detects human Tiki1/TRABD2A in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 901705 | |
| Western Blot 2 ug/mL | |
| Q86V40 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 129293 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction