Learn More
R&D Systems™ Human Thrombospondin-1 Quantikine ELISA Kit
Product Code. 18742004
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18742004
Brand: R&D Systems™ DTSP10
Human Thrombospondin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Thrombospondin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Thrombospondin-1 in cell culture supernates (25 ul), serum (10 ul), platelet-poor edta plasma (25 ul), platelet-poor heparin plasma (25 ul), saliva (50 ul), human milk (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 7.81 to 500 ng/mL
Assay Sensitivity 0.944 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA), Platelet-poor Plasma (EDTA, Heparin), Platelet-poor Plasma (Heparin), Saliva, Serum
Specificity Natural and Recombinant Human Thrombospondin-1
For Use With (Application) ELISA
Gene ID (Entrez) 7057
Gene Symbol THBS1
Quantity 1 Kit
Sample Volume Cell Culture Supernates (25uL),Serum (10uL),Platelet-poor EDTA Plasma (25uL),Platelet-poor Heparin Plasma (25uL),Saliva (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym THBS, THBS1, THBS-1, thrombospondin 1, Thrombospondin1, thrombospondin-1p180, TSP-1, TSP1thrombospondin-1, TSPTSP-1
Target Thrombospondin-1