missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Synaptojanin 2 (aa 1133-1209) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56784 (PA5-56784. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | O15056 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 8871 |
| Name | Human Synaptojanin 2 (aa 1133-1209) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI481647; inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; INPP5H; KIAA0348; LOW QUALITY PROTEIN: synaptojanin-2; mKIAA0348; phosphoinositide 5-phosphatase; SJ2; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; synaptojanin 2; synaptojanin-2; SYNJ2 |
| Common Name | Synaptojanin 2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction