missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STARD5 Full-length ORF (NP_871629.1, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16155023
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16155023 25 μg 25µg
16145023 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16155023 Supplier Abnova™ Supplier No. H00080765P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD5 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]

Sequence: MDPALAAQMSEAVAEKMLQYRRDTAGWKICREGNGVSVSWRPSVEFPGNLYRGEGIVYGTLEEVWDCVKPAVGGLRVKWDENVTGFEIIQSITDTLCVSRTSTPSAAMKLISPRDFVDLVLVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFHE

Specifications

Accession Number NP_871629.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80765
Molecular Weight (g/mol) 50.2kDa
Name STARD5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDPALAAQMSEAVAEKMLQYRRDTAGWKICREGNGVSVSWRPSVEFPGNLYRGEGIVYGTLEEVWDCVKPAVGGLRVKWDENVTGFEIIQSITDTLCVSRTSTPSAAMKLISPRDFVDLVLVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFHE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC10327
Common Name STARD5
Gene Symbol STARD5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.