Learn More
Gibco™ Human IL-15 Receptor alpha (soluble) Fc Recombinant Protein, PeproTech®
Description
Recombinant Human sIL-15 Receptor alpha Fc has a calculated molecular weight of 44.4 kDa protein and contains 408 amino acid residues, which consists of the extracellular domain of IL-15R alpha fused to the Fc portion of human IgG. Due to glycosylation, it migrates at an apparent molecular weight of 50-60 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q13261 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3601 |
| Molecular Weight (g/mol) | 44.4 kDa |
| Name | Human IL-15 Receptor alpha (soluble) Fc |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 20 μg |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.