missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin F1/PEDF Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 EUR - 1675.00 EUR
Specifications
| Antigen | Serpin F1/PEDF |
|---|---|
| Clone | 187003 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700837
![]() |
R&D Systems
MAB1177-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18798739
![]() |
R&D Systems
MAB1177-100 |
100 μg |
423.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713327
![]() |
R&D Systems
MAB1177-500 |
500 μg |
1675.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Serpin F1/PEDF | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| SERPINF1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 187003 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P36955 | |
| 5176 | |
| E. coli-derived recombinant human Serpin F1, Mouse myeloma cell line NS0-derived recombinant human Serpin F1, and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin F1 Asp44-Pro418 Accession # P36955 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Serpin F1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Serpin F1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title