Learn More
R&D Systems™ Human Semaphorin 3E, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 3239-S3B-025
Description
The Recombinant Human Semaphorin 3E Protein is derived from NS0. The Recombinant Human Semaphorin 3E Protein has been validated for the following applications: Bioactivity.Specifications
| O15041 | |
| 9723 | |
| 25μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E | |
| SEMA3E | |
| Unconjugated |
| Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Tween™ 20 with Trehalose. | |
| Observed M.W.: 85-100kDa, 66kDa and 25kDa, reducing conditions, Theoretical M.W.: 88kDa | |
| Mouse myeloma cell line,NS0-derived human Semaphorin 3E protein His24-Ser775 (Arg557Ala and Arg560Ala),with an N-terminal 10-His tag | |
| <1.0 EU per 1μg of the protein by the LAL method. | |
| Semaphorin 3E | |
| Measured by its ability to inhibit the proliferation of HUVEC human umbilical vein endothelial cells. Moriya, J. et al. (2010) Circ. Res. 106:391. The ED50 for this effect is typically 0.04-0.24μg/mL. Measured by its binding ability in a functional ELISA. When Recombinant Human Plexin D1 is coated at 5μg/mL, Recombinant Human Semaphorin 3E binds with an apparent Kd <0.4 nM. | |
| Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.