missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant PDGF-CC

Product Code. 17139741
Encompass_Preferred
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 17139741

Brand: STEMCELL Technologies 78168

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platelet-derived growth factor CC

  • The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer
  • PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ
  • It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src and PI3K/AKT
  • PDGF is a potent mitogen for cells of mesenchymal origin such as fibroblasts and vascular smooth muscle cells
  • PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues
  • PDGF-CC is secreted as a latent growth factor and requires activation by proteolytic processing
  • PDGF-CC binds to PDGFRα homodimers and PDGFRαβ heterodimers, but not to PDGFRβ homodimers

Specifications

Name PDGF, Platelet-derived growth factor
Quantity 10 μg
Cell Type Mesenchymal Cells, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Osteoblasts; Other
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.