missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant PDGF-CC
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer
- PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ
- It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src and PI3K/AKT
- PDGF is a potent mitogen for cells of mesenchymal origin such as fibroblasts and vascular smooth muscle cells
- PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues
- PDGF-CC is secreted as a latent growth factor and requires activation by proteolytic processing
- PDGF-CC binds to PDGFRα homodimers and PDGFRαβ heterodimers, but not to PDGFRβ homodimers
Specifications
Specifications
| Name | PDGF, Platelet-derived growth factor |
| Quantity | 50 μg |
| Cell Type | Mesenchymal Cells, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Osteoblasts; Other |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction