missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RalB Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3920
This item is not returnable.
View return policy
Description
RalB Monoclonal specifically detects RalB in Human samples. It is validated for Western Blot.
Specifications
| RalB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P11234 | |
| RALB | |
| E. coli-derived recombinant human RalB Met1-Leu206 Accession # P11234 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat RalB in Western blots. In Western blots, no cross-reactivity with recombinant human RalA is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 399417 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GTP binding protein, RAS-like protein B, ras-related protein Ral-B, v-ral simian leukemia viral oncogene homolog B (ras related; GTP bindingprotein) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5899 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction