missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PTH1R ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human PTH1R quantitates human PTH1R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PTH1R.
Parathyroid hormone receptor 1 is a member of the G-protein coupled receptor family. This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this receptor are known to be the cause of Jansen's metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), as well as enchodromatosis. Parathyroid hormone receptor 1 has been reported to be expressed in bone, breast, brain, colon, GI, kidney, liver, placenta, skin, umbilical cord, and uterus.
Specifications
Specifications
| Accession Number | MGC138426; MGC138452; Parathormone; Parathyrin; Parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; PTHR; PTHR1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor |
| Assay Range | 8.19 to 2,000 pg/mL |
| Assay Sensitivity | 8.2 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5745 |
| Gene Symbol | Pth1r |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction