Learn More
Abnova™ Human PRY Partial ORF (NP_004667.2, 48 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. It encodes a protein which has a low degree of similarity to protein tyrosine phosphatase, non-receptor type 13. Two nearly identical copies of this gene exist within a palindromic region. This record represents the more telomeric copy. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_004667.2 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 9081 |
| Molecular Weight (g/mol) | 36.74kDa |
| Name | PRY (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | EARRKKDLKDSFLWRYGKVGCISLPLREMTAWINPPQISEIFQGYHQRVHGADALSLQTNSLRSRLSSQCLGQSFLLRTLERGRGFRALGDICGHVHEED |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.